Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCTGATCAGACACAAAATCCTG[C/G]TGATGACATTTCCTCAATATTTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613127 | ||||||||||||||||||||
Literature Links: |
CHRDL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRDL2 - chordin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278473.2 | Intron | NP_001265402.1 | ||||
NM_001304390.1 | Intron | NP_001291319.1 | ||||
NM_001304391.1 | Intron | NP_001291320.1 | ||||
NM_001304415.1 | Intron | NP_001291344.1 | ||||
NM_001304416.1 | Intron | NP_001291345.1 | ||||
NM_001304417.1 | Intron | NP_001291346.1 | ||||
NM_015424.5 | Intron | NP_056239.3 |