Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTTCCACAGCTACAGTATACTT[A/G]GTTTGGAGTGGGGGATGGGGAATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606409 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITCH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITCH - itchy E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257137.2 | Intron | NP_001244066.1 | ||||
NM_001257138.2 | Intron | NP_001244067.1 | ||||
NM_001324197.1 | Intron | NP_001311126.1 | ||||
NM_001324198.1 | Intron | NP_001311127.1 | ||||
NM_031483.6 | Intron | NP_113671.3 | ||||
XM_005260570.1 | Intron | XP_005260627.1 | ||||
XM_011529079.1 | Intron | XP_011527381.1 | ||||
XM_017028089.1 | Intron | XP_016883578.1 | ||||
XM_017028090.1 | Intron | XP_016883579.1 | ||||
XM_017028091.1 | Intron | XP_016883580.1 | ||||
XM_017028092.1 | Intron | XP_016883581.1 |
LOC107985381 - uncharacterized LOC107985381 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017028185.1 | Intron | XP_016883674.1 |
Set Membership: |
HapMap |