Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGAGAGGCTGGAGAAGCCTGT[C/G]CCGTCAGAGAACCTTTGCCCCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616861 | ||||||||||||||||||||
Literature Links: |
SLC12A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC12A9 - solute carrier family 12 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267812.1 | Intron | NP_001254741.1 | ||||
NM_001267814.1 | Intron | NP_001254743.1 | ||||
NM_020246.3 | Intron | NP_064631.2 | ||||
XM_005250502.2 | Intron | XP_005250559.1 | ||||
XM_005250504.4 | Intron | XP_005250561.1 | ||||
XM_006716054.2 | Intron | XP_006716117.1 | ||||
XM_006716055.2 | Intron | XP_006716118.1 | ||||
XM_011516414.1 | Intron | XP_011514716.1 |