Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCTGATAGTGGCTTATGAAGGG[A/C]CCTTGTAAACTGACATGGCTGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609810 MIM: 604149 | ||||||||||||||||||||
Literature Links: |
PEG10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEG10 - paternally expressed 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040152.1 | Intron | NP_001035242.1 | ||||
NM_001172437.2 | Intron | NP_001165908.1 | ||||
NM_001172438.2 | Intron | NP_001165909.1 | ||||
NM_001184961.1 | Intron | NP_001171890.1 | ||||
NM_001184962.1 | Intron | NP_001171891.1 | ||||
NM_015068.3 | Intron | NP_055883.2 |
SGCE - sarcoglycan epsilon | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |