Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTGATATTTCTGCCAATTTTTG[G/T]TATATTGCTTTATCTTTTCTACTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151670 | ||||||||||||||||||||
Literature Links: |
LIPC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIPC - lipase C, hepatic type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000236.2 | Intron | NP_000227.2 | ||||
XM_005254372.1 | Intron | XP_005254429.1 | ||||
XM_005254374.4 | Intron | XP_005254431.2 | ||||
XM_006720502.3 | Intron | XP_006720565.1 | ||||
XM_017022176.1 | Intron | XP_016877665.1 |
LOC101928635 - uncharacterized LOC101928635 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011522293.2 | Intron | XP_011520595.1 |