Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTTGTTGTAAACTGGATGCTTT[C/T]AATAGAGAGAACCAGGGAAAAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607970 MIM: 614811 | ||||||||||||||||||||
Literature Links: |
GPR135 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR135 - G protein-coupled receptor 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022571.5 | Intron | NP_072093.2 | ||||
XM_017021596.1 | Intron | XP_016877085.1 | ||||
XM_017021597.1 | Intron | XP_016877086.1 | ||||
XM_017021598.1 | Intron | XP_016877087.1 | ||||
XM_017021599.1 | Intron | XP_016877088.1 |
L3HYPDH - trans-L-3-hydroxyproline dehydratase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |