Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAGGAACATTCACCAATTCTAT[A/C]AAAAATGATGTAGTTCTTTATCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606342 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIX4 - SIX homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017420.4 | 4504 | UTR 3 | NP_059116.3 | |||
XM_005267759.2 | 4504 | Intron | XP_005267816.1 |
Set Membership: |
HapMap |