Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATACTATATTGTAGCATCTAGT[A/G]TAAGCAAAGATTAATTTTTGTAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603348 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIF1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIF1A - hypoxia inducible factor 1 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243084.1 | Intron | NP_001230013.1 | ||||
NM_001530.3 | Intron | NP_001521.1 | ||||
NM_181054.2 | Intron | NP_851397.1 |
Set Membership: |
HapMap |