Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCCCCCACAGCCTTCCCCTCTC[A/G]TTCTTGCCATATGAAATGCACATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609868 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SPATA7 - spermatogenesis associated 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040428.3 | Intron | NP_001035518.1 | ||||
NM_018418.4 | Intron | NP_060888.2 | ||||
XM_005267851.1 | Intron | XP_005267908.1 | ||||
XM_005267852.1 | Intron | XP_005267909.1 | ||||
XM_005267854.1 | Intron | XP_005267911.1 | ||||
XM_005267855.1 | Intron | XP_005267912.1 | ||||
XM_006720204.1 | Intron | XP_006720267.1 | ||||
XM_006720205.1 | Intron | XP_006720268.1 | ||||
XM_011536951.1 | Intron | XP_011535253.1 | ||||
XM_011536952.1 | Intron | XP_011535254.1 | ||||
XM_011536953.1 | Intron | XP_011535255.1 | ||||
XM_017021452.1 | Intron | XP_016876941.1 | ||||
XM_017021453.1 | Intron | XP_016876942.1 | ||||
XM_017021454.1 | Intron | XP_016876943.1 | ||||
XM_017021455.1 | Intron | XP_016876944.1 | ||||
XM_017021456.1 | Intron | XP_016876945.1 | ||||
XM_017021457.1 | Intron | XP_016876946.1 |