Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612123 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPLA8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PNPLA8 - patatin like phospholipase domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256007.2 | Intron | NP_001242936.1 | ||||
NM_001256008.2 | Intron | NP_001242937.1 | ||||
NM_001256009.2 | Intron | NP_001242938.1 | ||||
NM_001256010.2 | Intron | NP_001242939.1 | ||||
NM_001256011.2 | Intron | NP_001242940.1 | ||||
NM_015723.4 | Intron | NP_056538.1 | ||||
XM_005250396.4 | Intron | XP_005250453.1 | ||||
XM_011516274.2 | Intron | XP_011514576.1 | ||||
XM_011516275.2 | Intron | XP_011514577.1 | ||||
XM_017012261.1 | Intron | XP_016867750.1 |
Set Membership: |
HapMap |