Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTGGTCCAAGATCTTTGGAAGA[C/T]TGAAGAAAGAGGATTACTTGAGCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603899 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF85 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF85 - zinc finger protein 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256171.1 | Intron | NP_001243100.1 | ||||
NM_001256172.1 | Intron | NP_001243101.1 | ||||
NM_001256173.1 | Intron | NP_001243102.1 | ||||
NM_003429.4 | Intron | NP_003420.2 | ||||
XM_011528263.2 | Intron | XP_011526565.1 | ||||
XM_011528264.1 | Intron | XP_011526566.1 | ||||
XM_017027234.1 | Intron | XP_016882723.1 |
Set Membership: |
HapMap |