Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAATGCCCACACAGTTGTGCAT[C/T]TTTTTTTGTTTTTCCCCTGACTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604992 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SRL - sarcalumenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098814.1 | 3809 | UTR 3 | NP_001092284.1 | |||
NM_001323667.1 | 3809 | UTR 3 | NP_001310596.1 | |||
NM_001323668.1 | 3809 | UTR 3 | NP_001310597.1 | |||
XM_017023527.1 | 3809 | UTR 3 | XP_016879016.1 | |||
XM_017023528.1 | 3809 | UTR 3 | XP_016879017.1 | |||
XM_017023529.1 | 3809 | UTR 3 | XP_016879018.1 |
Set Membership: |
HapMap |