Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614666 MIM: 610998 MIM: 611118 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC78 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC78 - coiled-coil domain containing 78 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM173A - family with sequence similarity 173 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAGHL - hydroxyacylglutathione hydrolase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290137.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | NP_001277066.1 | |
NM_001290139.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | NP_001277068.1 | |
NM_001323635.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | NP_001310564.1 | |
NM_001323636.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | NP_001310565.1 | |
NM_032304.3 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | NP_115680.1 | |
XM_005255631.4 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | XP_005255688.1 | |
XM_005255632.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | XP_005255689.1 | |
XM_011522711.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | XP_011521013.1 | |
XM_011522712.1 | 1020 | Silent Mutation | ACA,ACG | T,T 174 | XP_011521014.1 | |
XM_017023773.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | XP_016879262.1 | |
XM_017023774.1 | 1020 | Silent Mutation | ACA,ACG | T,T 175 | XP_016879263.1 |
METRN - meteorin, glial cell differentiation regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NARFL - nuclear prelamin A recognition factor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304799.1 | 1020 | Intron | NP_001291728.1 | |||
NM_022493.2 | 1020 | Intron | NP_071938.1 |
Set Membership: |
HapMap |