Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGAGAGAAATCTTTGATTTCT[A/G]TACCTTAGTCTATATTCCTTAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602207 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAB18 - RAB18, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256410.1 | Intron | NP_001243339.1 | ||||
NM_001256411.1 | Intron | NP_001243340.1 | ||||
NM_001256412.1 | Intron | NP_001243341.1 | ||||
NM_001256415.1 | Intron | NP_001243344.1 | ||||
NM_021252.4 | Intron | NP_067075.1 |