Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGGACTTAAGCAAACTCGTTTC[A/G]CTTCTCTAAGCCTTCCTTTATTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606503 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUV39H2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUV39H2 - suppressor of variegation 3-9 homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193424.1 | Intron | NP_001180353.1 | ||||
NM_001193425.1 | Intron | NP_001180354.1 | ||||
NM_001193426.1 | Intron | NP_001180355.1 | ||||
NM_001193427.1 | Intron | NP_001180356.1 | ||||
NM_024670.3 | Intron | NP_078946.1 | ||||
XM_006717503.3 | Intron | XP_006717566.1 | ||||
XM_011519662.2 | Intron | XP_011517964.1 | ||||
XM_017016637.1 | Intron | XP_016872126.1 |
Set Membership: |
HapMap |