Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCCAGTTACACATGCATGAAGG[C/T]TGCCCTGCCCACTGGTTCCTGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600902 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPHS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEPHS1 - selenophosphate synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195602.1 | 3080 | Intron | NP_001182531.1 | |||
NM_001195604.1 | 3080 | Intron | NP_001182533.1 | |||
NM_012247.4 | 3080 | Intron | NP_036379.2 | |||
XM_017015942.1 | 3080 | UTR 3 | XP_016871431.1 | |||
XM_017015943.1 | 3080 | UTR 3 | XP_016871432.1 | |||
XM_017015944.1 | 3080 | UTR 3 | XP_016871433.1 | |||
XM_017015945.1 | 3080 | Intron | XP_016871434.1 |
Set Membership: |
HapMap |