Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGTGCTCAGGCCTAGTCTCACCT[C/T]GGCCCAGCTGGGTGCAGGGCCCAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNU6-6P PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNU6-6P - RNA, U6 small nuclear 6, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCMA - upper zone of growth plate and cartilage matrix associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303118.1 | Intron | NP_001290047.1 | ||||
NM_001303119.1 | Intron | NP_001290048.1 | ||||
NM_145314.2 | Intron | NP_660357.2 |
Set Membership: |
HapMap |