Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGTTGGAATTTTGGCGGGTTCA[A/G]CTGTGAACGAAGAAGGCGTCCCGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609357 | |||||||||||||||||||||||
Literature Links: |
MCM10 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MCM10 - minichromosome maintenance 10 replication initiation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018518.4 | 23 | UTR 5 | NP_060988.3 | |||
NM_182751.2 | 23 | UTR 5 | NP_877428.1 | |||
XM_011519538.2 | 23 | Intron | XP_011517840.1 |