Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATATCTTATAAAGCTAATAGA[A/G]AAAATCTACAAAGTAACTTTTCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf34 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C8orf34 - chromosome 8 open reading frame 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195639.1 | Intron | NP_001182568.1 | ||||
NM_052958.2 | Intron | NP_443190.2 | ||||
XM_011517445.2 | Intron | XP_011515747.1 | ||||
XM_011517446.2 | Intron | XP_011515748.1 | ||||
XM_011517448.2 | Intron | XP_011515750.1 | ||||
XM_011517449.2 | Intron | XP_011515751.1 | ||||
XM_011517450.2 | Intron | XP_011515752.1 | ||||
XM_017013019.1 | Intron | XP_016868508.1 |
C8orf34-AS1 - C8orf34 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |