Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATACTGAACCCATTTTGTGCCCA[C/G]GGATATTGTGATGACAAAACAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603088 | ||||||||||||||||||||||||||||||||
Literature Links: |
FKBP15 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
FKBP15 - FK506 binding protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015258.1 | Intron | NP_056073.1 | ||||
XM_006717018.2 | Intron | XP_006717081.1 | ||||
XM_006717019.1 | Intron | XP_006717082.1 | ||||
XM_017014552.1 | Intron | XP_016870041.1 |
SLC31A2 - solute carrier family 31 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |