Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAAAAGAGTATGCCAAATTATT[C/T]TGCCCAAAAGAGAATTTCACCCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609720 | ||||||||||||||||||||
Literature Links: |
CRB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRB2 - crumbs 2, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173689.6 | Intron | NP_775960.4 | ||||
XM_005251934.2 | Intron | XP_005251991.1 | ||||
XM_011518556.2 | Intron | XP_011516858.1 | ||||
XM_011518557.2 | Intron | XP_011516859.1 | ||||
XM_011518558.2 | Intron | XP_011516860.1 |