Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCACTATATTGACTTTAACACC[A/C]CAGATTAGTTTTGTCCTTTGTGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604602 | ||||||||||||||||||||
Literature Links: |
MRRF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRRF - mitochondrial ribosome recycling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173512.1 | Intron | NP_001166983.1 | ||||
NM_138777.3 | Intron | NP_620132.1 | ||||
NM_199177.2 | Intron | NP_954646.1 | ||||
XM_005252302.3 | Intron | XP_005252359.1 | ||||
XM_011519183.2 | Intron | XP_011517485.1 | ||||
XM_011519184.2 | Intron | XP_011517486.1 | ||||
XM_011519185.2 | Intron | XP_011517487.1 | ||||
XM_011519186.1 | Intron | XP_011517488.1 | ||||
XM_011519187.2 | Intron | XP_011517489.1 | ||||
XM_017015286.1 | Intron | XP_016870775.1 | ||||
XM_017015287.1 | Intron | XP_016870776.1 |
RBM18 - RNA binding motif protein 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |