Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGACATCCTGGGGTCTTGAGA[A/G]TCCTGGAGAGACTGCTCCTCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615365 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK8 - adenylate kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317958.1 | Intron | NP_001304887.1 | ||||
NM_001317959.1 | Intron | NP_001304888.1 | ||||
NM_152572.2 | Intron | NP_689785.1 | ||||
XM_005272169.2 | Intron | XP_005272226.1 | ||||
XM_006716965.2 | Intron | XP_006717028.1 | ||||
XM_011518277.2 | Intron | XP_011516579.1 | ||||
XM_011518278.2 | Intron | XP_011516580.1 | ||||
XM_017014308.1 | Intron | XP_016869797.1 |
Set Membership: |
HapMap |