Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGGTGCCTTCAGAATTCTGAAA[A/G]GGCCCTGGATGGTATCGGGTAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604383 MIM: 605284 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFI1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GFI1B - growth factor independent 1B transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135031.1 | Intron | NP_001128503.1 | ||||
NM_004188.6 | Intron | NP_004179.3 | ||||
XM_006717297.3 | Intron | XP_006717360.1 | ||||
XM_011519068.2 | Intron | XP_011517370.1 | ||||
XM_011519069.2 | Intron | XP_011517371.1 | ||||
XM_011519070.2 | Intron | XP_011517372.1 | ||||
XM_017015175.1 | Intron | XP_016870664.1 | ||||
XM_017015176.1 | Intron | XP_016870665.1 |
MIR548AW - microRNA 548aw | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSC1 - tuberous sclerosis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000368.4 | Intron | NP_000359.1 | ||||
NM_001162426.1 | Intron | NP_001155898.1 | ||||
NM_001162427.1 | Intron | NP_001155899.1 | ||||
XM_005272211.1 | Intron | XP_005272268.1 | ||||
XM_006717271.1 | Intron | XP_006717334.1 | ||||
XM_011518979.2 | Intron | XP_011517281.1 | ||||
XM_017015096.1 | Intron | XP_016870585.1 | ||||
XM_017015097.1 | Intron | XP_016870586.1 | ||||
XM_017015098.1 | Intron | XP_016870587.1 | ||||
XM_017015099.1 | Intron | XP_016870588.1 | ||||
XM_017015100.1 | Intron | XP_016870589.1 | ||||
XM_017015101.1 | Intron | XP_016870590.1 |
Set Membership: |
HapMap |