Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATCTAAGAAATCCCAGTCGGT[A/G]GTTTGTCCATATACCATGATTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614085 MIM: 604730 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHNO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RHNO1 - RAD9-HUS1-RAD1 interacting nuclear orphan 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TULP3 - tubby like protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160408.1 | Intron | NP_001153880.1 | ||||
NM_003324.4 | Intron | NP_003315.2 |
Set Membership: |
HapMap |