Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAATGAACTGGATAGGGACCAAA[C/G]GATTTCCTTCACATATACGGGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611368 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILDR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ILDR2 - immunoglobulin like domain containing receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199351.2 | Intron | NP_955383.1 | ||||
XM_006711314.3 | Intron | XP_006711377.1 | ||||
XM_017001252.1 | Intron | XP_016856741.1 | ||||
XM_017001253.1 | Intron | XP_016856742.1 | ||||
XM_017001254.1 | Intron | XP_016856743.1 | ||||
XM_017001255.1 | Intron | XP_016856744.1 | ||||
XM_017001256.1 | Intron | XP_016856745.1 | ||||
XM_017001257.1 | Intron | XP_016856746.1 | ||||
XM_017001258.1 | Intron | XP_016856747.1 |
MAEL - maelstrom spermatogenic transposon silencer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286377.1 | Intron | NP_001273306.1 | ||||
NM_001286378.1 | Intron | NP_001273307.1 | ||||
NM_032858.2 | Intron | NP_116247.1 | ||||
XM_006711583.1 | Intron | XP_006711646.1 | ||||
XM_011510067.1 | Intron | XP_011508369.1 | ||||
XM_011510068.1 | Intron | XP_011508370.1 | ||||
XM_017002602.1 | Intron | XP_016858091.1 | ||||
XM_017002603.1 | Intron | XP_016858092.1 |
Set Membership: |
HapMap |