Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTCACCTCCTCCCCACTCTGG[A/G]ACTCTGGGATCCCTCTGAACTGCTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600298 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMX1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LMX1A - LIM homeobox transcription factor 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174069.1 | 2156 | UTR 3 | NP_001167540.1 | |||
NM_177398.3 | 2156 | UTR 3 | NP_796372.1 | |||
XM_011509538.2 | 2156 | UTR 3 | XP_011507840.1 | |||
XM_011509540.2 | 2156 | Intron | XP_011507842.1 |
Set Membership: |
HapMap |