Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGATCCTGAGAGTAGGAAAGGG[A/T]ACACTGAGTCTTTTCAGTTGAAGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131210 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SELE PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SELE - selectin E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000450.2 | 2196 | UTR 3 | NP_000441.2 |