Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGGTAATTATATGGAAAGAAC[G/T]GTGGCTAACCAGACTCTTTGATTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL11B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
METTL11B - methyltransferase like 11B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136107.1 | Intron | NP_001129579.1 | ||||
XM_011509232.2 | Intron | XP_011507534.1 | ||||
XM_011509233.2 | Intron | XP_011507535.1 | ||||
XM_011509234.2 | Intron | XP_011507536.1 |
MIR3119-1 - microRNA 3119-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3119-2 - microRNA 3119-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |