Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTCATATATACGATTTCAAAAT[A/T]AAGTCAAAAATTTTAATACATCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610044 | ||||||||||||||||||||
Literature Links: |
KCNT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNT2 - potassium sodium-activated channel subfamily T member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287819.1 | 3597 | UTR 3 | NP_001274748.1 | |||
NM_001287820.1 | 3597 | UTR 3 | NP_001274749.1 | |||
NM_198503.3 | 3597 | UTR 3 | NP_940905.2 | |||
XM_006711294.2 | 3597 | UTR 3 | XP_006711357.1 | |||
XM_006711295.2 | 3597 | UTR 3 | XP_006711358.1 | |||
XM_011509482.2 | 3597 | UTR 3 | XP_011507784.1 | |||
XM_011509483.2 | 3597 | Intron | XP_011507785.1 | |||
XM_017001178.1 | 3597 | Intron | XP_016856667.1 | |||
XM_017001179.1 | 3597 | Intron | XP_016856668.1 | |||
XM_017001180.1 | 3597 | Intron | XP_016856669.1 | |||
XM_017001181.1 | 3597 | UTR 3 | XP_016856670.1 | |||
XM_017001182.1 | 3597 | UTR 3 | XP_016856671.1 | |||
XM_017001183.1 | 3597 | UTR 3 | XP_016856672.1 | |||
XM_017001184.1 | 3597 | UTR 3 | XP_016856673.1 | |||
XM_017001185.1 | 3597 | Intron | XP_016856674.1 |