Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTGCCTTTGAGGGCAATGACAG[A/C]CAATGTTCTCGCTTTAAGAAATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIQK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIQK - triple QxxK/R motif containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171795.1 | Intron | NP_001165266.1 | ||||
NM_001171796.1 | Intron | NP_001165267.1 | ||||
NM_001171797.1 | Intron | NP_001165268.1 | ||||
NM_001171798.1 | Intron | NP_001165269.1 | ||||
NM_001171799.1 | Intron | NP_001165270.1 | ||||
NM_001191035.1 | Intron | NP_001177964.1 | ||||
NM_001191036.1 | Intron | NP_001177965.1 | ||||
XM_017013347.1 | Intron | XP_016868836.1 | ||||
XM_017013348.1 | Intron | XP_016868837.1 | ||||
XM_017013349.1 | Intron | XP_016868838.1 | ||||
XM_017013350.1 | Intron | XP_016868839.1 |