Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTATCAGGATATTATTTGTTT[A/G]TTTTGGGGTAATATACAAATAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609884 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM67 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM67 - transmembrane protein 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142301.1 | Intron | NP_001135773.1 | ||||
NM_153704.5 | Intron | NP_714915.3 | ||||
XM_006716686.3 | Intron | XP_006716749.1 | ||||
XM_011517363.2 | Intron | XP_011515665.1 |