Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTGTATAAACAGCAACCCAGG[A/G]ATCTTCCCAGGAACCTCAGCCATCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603398 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WISP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WISP1 - WNT1 inducible signaling pathway protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204869.1 | Intron | NP_001191798.1 | ||||
NM_001204870.1 | Intron | NP_001191799.1 | ||||
NM_003882.3 | Intron | NP_003873.1 | ||||
NM_080838.2 | Intron | NP_543028.1 |
Set Membership: |
HapMap |