Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCCGGGGGTGACTTCAAACAG[A/C]CGCAAGATTGGGCTCTGGGGAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRR15 - proline rich 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175887.2 | 1046 | Intron | NP_787083.1 | |||
XM_005249653.3 | 1046 | UTR 5 | XP_005249710.1 | |||
XM_011515198.1 | 1046 | UTR 5 | XP_011513500.1 | |||
XM_011515199.1 | 1046 | Intron | XP_011513501.1 | |||
XM_017011842.1 | 1046 | Intron | XP_016867331.1 |