Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGTTTACAAAGCATGAGAAATA[C/T]AGCCACAGTATGTTGTAAACTGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608202 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK5RAP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDK5RAP3 - CDK5 regulatory subunit associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278197.1 | Intron | NP_001265126.1 | ||||
NM_001278198.1 | Intron | NP_001265127.1 | ||||
NM_001278216.1 | Intron | NP_001265145.1 | ||||
NM_001278217.1 | Intron | NP_001265146.1 | ||||
NM_176096.2 | Intron | NP_788276.1 | ||||
XM_011525297.1 | Intron | XP_011523599.1 | ||||
XM_017025164.1 | Intron | XP_016880653.1 | ||||
XM_017025165.1 | Intron | XP_016880654.1 | ||||
XM_017025166.1 | Intron | XP_016880655.1 |