Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTGCTGCCCACCACTGCTCAAA[G/T]TCCTTCTCCACGCATCAACCCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609785 | ||||||||||||||||||||
Literature Links: |
TFCP2L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TFCP2L1 - transcription factor CP2-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014553.2 | 7439 | UTR 3 | NP_055368.1 | |||
XM_017003902.1 | 7439 | UTR 3 | XP_016859391.1 | |||
XM_017003903.1 | 7439 | UTR 3 | XP_016859392.1 | |||
XM_017003904.1 | 7439 | Intron | XP_016859393.1 | |||
XM_017003905.1 | 7439 | Intron | XP_016859394.1 |