Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTAGTTTATATGCCTGTAACAC[A/G]TATCACTGTAATTCTGGGTCCTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IWS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IWS1 - IWS1, SUPT6H interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017969.2 | Intron | NP_060439.2 | ||||
XM_005263707.3 | Intron | XP_005263764.1 | ||||
XM_006712626.3 | Intron | XP_006712689.1 | ||||
XM_011511450.1 | Intron | XP_011509752.1 | ||||
XM_011511452.1 | Intron | XP_011509754.1 | ||||
XM_017004467.1 | Intron | XP_016859956.1 | ||||
XM_017004468.1 | Intron | XP_016859957.1 |