Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGCAGTTACTGTTAATATTTCC[C/T]TTTTGTTAGAGAGCTGGTTTTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613438 | ||||||||||||||||||||
Literature Links: |
FCHO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCHO2 - FCH domain only 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146032.1 | Intron | NP_001139504.1 | ||||
NM_138782.2 | Intron | NP_620137.2 | ||||
XM_017009016.1 | Intron | XP_016864505.1 | ||||
XM_017009017.1 | Intron | XP_016864506.1 | ||||
XM_017009018.1 | Intron | XP_016864507.1 | ||||
XM_017009019.1 | Intron | XP_016864508.1 | ||||
XM_017009020.1 | Intron | XP_016864509.1 | ||||
XM_017009021.1 | Intron | XP_016864510.1 | ||||
XM_017009022.1 | Intron | XP_016864511.1 | ||||
XM_017009023.1 | Intron | XP_016864512.1 |