Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601953 MIM: 139150 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNH PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCNH - cyclin H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199189.1 | Intron | NP_001186118.1 | ||||
NM_001239.3 | Intron | NP_001230.1 | ||||
XM_005248627.3 | Intron | XP_005248684.1 | ||||
XM_005248629.4 | Intron | XP_005248686.1 | ||||
XM_011543706.2 | Intron | XP_011542008.1 | ||||
XM_017010014.1 | Intron | XP_016865503.1 |
RASA1 - RAS p21 protein activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |