Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAAAAAAGAAAGCCAGGGCCCA[A/G]GAGTCCCAATCCACCACTCACCAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603679 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2N PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBE2N - ubiquitin conjugating enzyme E2 N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003348.3 | Intron | NP_003339.1 | ||||
XM_011538719.2 | Intron | XP_011537021.1 | ||||
XM_017019918.1 | Intron | XP_016875407.1 |
Set Membership: |
HapMap |