Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCCTATTCACTGTCTTGGTGGT[A/T]CCCTTGGAAGTACAAAAGTTTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EML6 - echinoderm microtubule associated protein like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039753.2 | Intron | NP_001034842.2 | ||||
XM_017004098.1 | Intron | XP_016859587.1 | ||||
XM_017004099.1 | Intron | XP_016859588.1 | ||||
XM_017004100.1 | Intron | XP_016859589.1 | ||||
XM_017004101.1 | Intron | XP_016859590.1 | ||||
XM_017004102.1 | Intron | XP_016859591.1 |