Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCCAATAGTATATATTTTTAGA[G/T]ATATTCATTTATATATTTTTAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP36 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFAP36 - cilia and flagella associated protein 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282761.1 | Intron | NP_001269690.1 | ||||
NM_080667.6 | Intron | NP_542398.3 | ||||
XM_011532499.1 | Intron | XP_011530801.1 | ||||
XM_017003254.1 | Intron | XP_016858743.1 | ||||
XM_017003255.1 | Intron | XP_016858744.1 |