Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATGTACAAATATGTACAGTCAT[C/T]GTCTAAAACAACATTGCCAGGACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM179A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM179A - family with sequence similarity 179 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321538.1 | Intron | NP_001308467.1 | ||||
NM_001321539.1 | Intron | NP_001308468.1 | ||||
NM_199280.2 | Intron | NP_954974.2 | ||||
XM_011532628.2 | Intron | XP_011530930.1 | ||||
XM_011532631.1 | Intron | XP_011530933.1 | ||||
XM_017003507.1 | Intron | XP_016858996.1 | ||||
XM_017003508.1 | Intron | XP_016858997.1 | ||||
XM_017003509.1 | Intron | XP_016858998.1 | ||||
XM_017003510.1 | Intron | XP_016858999.1 | ||||
XM_017003511.1 | Intron | XP_016859000.1 | ||||
XM_017003512.1 | Intron | XP_016859001.1 | ||||
XM_017003513.1 | Intron | XP_016859002.1 | ||||
XM_017003514.1 | Intron | XP_016859003.1 | ||||
XM_017003515.1 | Intron | XP_016859004.1 | ||||
XM_017003516.1 | Intron | XP_016859005.1 |
Set Membership: |
HapMap |