Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAACCACTTATTTTATTCTGTCT[C/T]TTACAAGAAATCCTGAAGAAACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603282 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF204P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF204P - zinc finger protein 204, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |