Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAATTGTTTTGAGGTGTCTGGA[A/C]CCATGCCCATAGGCAGACTTAATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600289 | ||||||||||||||||||||
Literature Links: |
MAPK14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPK14 - mitogen-activated protein kinase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001315.2 | Intron | NP_001306.1 | ||||
NM_139012.2 | Intron | NP_620581.1 | ||||
NM_139013.2 | Intron | NP_620582.1 | ||||
NM_139014.2 | Intron | NP_620583.1 | ||||
XM_006714998.3 | Intron | XP_006715061.1 | ||||
XM_011514310.2 | Intron | XP_011512612.2 | ||||
XM_017010299.1 | Intron | XP_016865788.1 | ||||
XM_017010300.1 | Intron | XP_016865789.1 | ||||
XM_017010301.1 | Intron | XP_016865790.1 | ||||
XM_017010302.1 | Intron | XP_016865791.1 | ||||
XM_017010303.1 | Intron | XP_016865792.1 | ||||
XM_017010304.1 | Intron | XP_016865793.1 |