Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTACTCTGACCCATCGTCGAAC[C/G]TAAGCCAGTTATATTGAGTTCAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611827 | ||||||||||||||||||||
Literature Links: |
MRPL14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL14 - mitochondrial ribosomal protein L14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM63B - transmembrane protein 63B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318792.1 | Intron | NP_001305721.1 | ||||
NM_018426.2 | Intron | NP_060896.1 | ||||
XM_005249211.3 | Intron | XP_005249268.1 | ||||
XM_005249213.4 | Intron | XP_005249270.1 | ||||
XM_005249214.4 | Intron | XP_005249271.1 | ||||
XM_005249215.4 | Intron | XP_005249272.1 | ||||
XM_005249217.1 | Intron | XP_005249274.1 | ||||
XM_006715135.1 | Intron | XP_006715198.1 | ||||
XM_017010999.1 | Intron | XP_016866488.1 | ||||
XM_017011000.1 | Intron | XP_016866489.1 | ||||
XM_017011001.1 | Intron | XP_016866490.1 | ||||
XM_017011002.1 | Intron | XP_016866491.1 |