Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCTTAAATTTGGGACTGTGTGA[A/G]AATCAAGTTTCTAAACCCTCTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605994 MIM: 613725 | ||||||||||||||||||||
Literature Links: |
CYP39A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP39A1 - cytochrome P450 family 39 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A27 - solute carrier family 25 member 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204051.1 | 23 | Intron | NP_001190980.1 | |||
NM_001204052.1 | 23 | Intron | NP_001190981.1 | |||
NM_004277.4 | 23 | Intron | NP_004268.3 | |||
XM_005249483.2 | 23 | Intron | XP_005249540.1 | |||
XM_017011512.1 | 23 | UTR 5 | XP_016867001.1 |