Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGACCGAGCAGCCTCCTCCTC[C/T]TAGGATGACCTCACCCTCCAGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606676 | ||||||||||||||||||||
Literature Links: |
TRPV2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRPV2 - transient receptor potential cation channel subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016113.4 | 395 | UTR 5 | NP_057197.2 | |||
XM_005256676.1 | 395 | UTR 5 | XP_005256733.1 | |||
XM_005256677.1 | 395 | UTR 5 | XP_005256734.1 | |||
XM_005256678.4 | 395 | UTR 5 | XP_005256735.1 | |||
XM_006721541.3 | 395 | UTR 5 | XP_006721604.1 | |||
XM_006721543.3 | 395 | UTR 5 | XP_006721606.1 | |||
XM_011523922.1 | 395 | UTR 5 | XP_011522224.1 | |||
XM_011523923.1 | 395 | UTR 5 | XP_011522225.1 | |||
XM_011523925.2 | 395 | Intron | XP_011522227.1 | |||
XM_017024730.1 | 395 | UTR 5 | XP_016880219.1 | |||
XM_017024731.1 | 395 | UTR 5 | XP_016880220.1 | |||
XM_017024732.1 | 395 | UTR 5 | XP_016880221.1 |