Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1211 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA1211 - KIAA1211 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020722.1 | Intron | NP_065773.1 | ||||
XM_005265752.2 | Intron | XP_005265809.1 | ||||
XM_005265755.2 | Intron | XP_005265812.1 | ||||
XM_005265756.2 | Intron | XP_005265813.1 | ||||
XM_006714045.2 | Intron | XP_006714108.1 | ||||
XM_011534397.2 | Intron | XP_011532699.1 | ||||
XM_017008473.1 | Intron | XP_016863962.1 | ||||
XM_017008474.1 | Intron | XP_016863963.1 |