Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612620 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RASSF6 - Ras association domain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270391.1 | 4033 | UTR 3 | NP_001257320.1 | |||
NM_001270392.1 | 4033 | UTR 3 | NP_001257321.1 | |||
NM_177532.4 | 4033 | UTR 3 | NP_803876.1 | |||
NM_201431.2 | 4033 | UTR 3 | NP_958834.1 | |||
XM_011531687.2 | 4033 | Intron | XP_011529989.1 | |||
XM_011531688.1 | 4033 | UTR 3 | XP_011529990.1 | |||
XM_011531689.2 | 4033 | UTR 3 | XP_011529991.1 | |||
XM_011531690.2 | 4033 | UTR 3 | XP_011529992.1 | |||
XM_011531691.1 | 4033 | UTR 3 | XP_011529993.1 | |||
XM_017007837.1 | 4033 | UTR 3 | XP_016863326.1 | |||
XM_017007838.1 | 4033 | Intron | XP_016863327.1 |
Set Membership: |
HapMap |